Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitic acid (Standard) | 57-10-3 | 256.42 | 100 MG
SDP

Catalog No. 5000416438
Encompass_Preferred

Palmitic acid (Standard) is provided as an analytical standard for Palmitic acid, intended specifically for research and analytical applications. It is a long-chain saturated fatty acid naturally present in both animal and plant sources. This compound has been observed to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.

  • Purity: 98.31%
  • Application: Serves as a reference standard for assays and is commonly utilized in qualitative, quantitative, and methodological research experiments, particularly with techniques such as HPLC, GC, and MS.
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Plants (other families), Endogenous metabolite, Human Gut Microbiota Metabolites, Animals, Microorganisms.
  • Formula: C₁₆H₃₂O₂

Catalog No. 50-004-16438 Supplier Medchemexpress LLC Supplier No. HYN0830R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitic acid (Standard) is provided as an analytical standard for Palmitic acid, intended specifically for research and analytical applications. It is a long-chain saturated fatty acid naturally present in both animal and plant sources. This compound has been observed to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.

  • Purity: 98.31%
  • Application: Serves as a reference standard for assays and is commonly utilized in qualitative, quantitative, and methodological research experiments, particularly with techniques such as HPLC, GC, and MS.
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Plants (other families), Endogenous metabolite, Human Gut Microbiota Metabolites, Animals, Microorganisms.
  • Formula: C₁₆H₃₂O₂

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.