Learn More
Medchemexpress LLC Palmitic acid (Standard) | 57-10-3 | MFCD00002747 | 256.42 | 50 MG
SDP
Catalog No. 5000416957
Encompass_Preferred
Palmitic acid (Standard) is the analytical standard of Palmitic acid. This product is intended for research and analytical applications. It is a long-chain saturated fatty acid commonly found in both animals and plants. The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Induces the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
Catalog No. 50-004-16957 Supplier Medchemexpress LLC Supplier No. HYN0830R50MG
Palmitic acid (Standard) is the analytical standard of Palmitic acid. This product is intended for research and analytical applications. It is a long-chain saturated fatty acid commonly found in both animals and plants. The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Induces the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.