Learn More
Cayman Chemical PalmItIc AcId13CC2 labeled 5mg
SDP
Catalog No. 502938460
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-293-8460 Supplier Cayman Chemical Supplier No. 289165 MG
Add to Cart
Add to Cart
An internal standard for the quantification of palmitic acid by GC- or LC-MS