Learn More
Medchemexpress LLC Palmitic anhydride 50g
SDP
Catalog No. 5000206547
Encompass_Preferred
Palmitic anhydride a saturated fatty acid found in many natural sources such as animal fats and vegetable oils is commonly used as a reagent in organic synthesis to introduce palmitoyl(hexadecanoyl) functional groups into other organic molecules moreover Palmitic anhydride has been Used in the production of surfactants lubricants and plasticizers as well as in the preparation of cosmetics and pharmaceuticals
Catalog No. 50-002-06547 Supplier Medchemexpress LLC Supplier No. HYW09318350G
Palmitic anhydride a saturated fatty acid found in many natural sources such as animal fats and vegetable oils is commonly used as a reagent in organic synthesis to introduce palmitoyl(hexadecanoyl) functional groups into other organic molecules moreover Palmitic anhydride has been Used in the production of surfactants lubricants and plasticizers as well as in the preparation of cosmetics and pharmaceuticals