Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitic-d31 acid, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014136 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4136 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4136 Supplier MilliporeSigma Supelco Supplier No. 6827725MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 39756-30-4
Shipping Condition Ambient
Physical Form Neat
Linear Formula CD3(CD2)14 CO2 H
MDL Number MFCD00053335
Melting Point 58°C to 60°C; 61°C to 64°C
Synonym Palmitic acid-d31; Hexadecanoic-d31 acid
Type Analytical Standard
Quantity 25 mg
Formula Weight 287.62
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.