Learn More
Cayman Chemical Palmitoleic Acid, 373-49-9, 100 mg
SDP
Supplier:  Cayman Chemical 10009871100
Encompass_Preferred
Molecular Formula C16H30O2, Purity ≥99%, Formula Weight 254.4, Synonyms: 9-cis-Hexadecenoic Acid, n-7 Palmitoleate, Palmitoleate, cis-Palmitoleic Acid, (9Z)-hexadecenoic acid; An -7 monounsaturated fatty acid that is a common constituent of the triglycerides of human adipose tissue; raises LDL cholesterol and lowers HDL cholesterol much like that of a saturated fatty acid, even when dietary intake of cholesterol is maintained at a low level.
Catalog No. NC9901001