Learn More
Medchemexpress LLC Palmitoleic acid-13C16 | 2483735-57-3 | 99.20% | 270.29 | 1 MG
SDP
Catalog No. 5000453819
Encompass_Preferred
Palmitoleic acid-13C16 is the 13C labeled Palmitoleic acid. Palmitoleic acid, a composition of fatty acid, is implicated in the prevention of death from cerebrovascular disorders in SHRSP rats.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-53819 Supplier Medchemexpress LLC Supplier No. HYW011873S21MG
Palmitoleic acid-13C16 is the 13C labeled Palmitoleic acid. Palmitoleic acid, a composition of fatty acid, is implicated in the prevention of death from cerebrovascular disorders in SHRSP rats.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.