Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitoleic acid | 373-49-9 | 99.8% | 254.41 | 1 ML
SDP

Catalog No. 5000453929
Encompass_Preferred

Palmitoleic acid, a fatty acid, is implicated in the prevention of death from cerebrovascular disorders in SHRSP rats. It is intended for research use only. This endogenous metabolite is also found in human milk and has shown to significantly improve survival rates with a concomitant reduction in stroke incidence in SHRSP rats, even with excess NaCl intake.

  • Intended for research use only
  • High purity of 99.84%
  • Molecular weight is 254.41
  • Appears as a colorless to light yellow liquid
  • Provided as a 10 mM solution in DMSO
  • Long-term storage at -20°C (pure) or -80°C (in solvent)

Catalog No. 50-004-53929 Supplier Medchemexpress LLC Supplier No. HYW01187310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitoleic acid, a fatty acid, is implicated in the prevention of death from cerebrovascular disorders in SHRSP rats. It is intended for research use only. This endogenous metabolite is also found in human milk and has shown to significantly improve survival rates with a concomitant reduction in stroke incidence in SHRSP rats, even with excess NaCl intake.

  • Intended for research use only
  • High purity of 99.84%
  • Molecular weight is 254.41
  • Appears as a colorless to light yellow liquid
  • Provided as a 10 mM solution in DMSO
  • Long-term storage at -20°C (pure) or -80°C (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.