Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitoleic acid | 373-49-9 | 99.8% | 254.41 | 50 MG
SDP

Catalog No. 5000453707
Encompass_Preferred

Palmitoleic acid is a fatty acid involved in preventing death from cerebrovascular disorders in SHRSP rats. It is an endogenous metabolite found in human milk. Studies have shown that a diet containing 1% Palmitoleic acid significantly improves the survival rate and reduces the incidence of stroke in SHRSP rats, even with excess NaCl intake.

  • Endogenous metabolite
  • Purity of 99.84%
  • Formula C16H30O2
  • CAS number 373-49-9
  • Appears as a colorless to light yellow liquid
  • Significant improvement in survival rate and reduced incidence of stroke in SHRSP rats

Catalog No. 50-004-53707 Supplier Medchemexpress LLC Supplier No. HYW01187350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitoleic acid is a fatty acid involved in preventing death from cerebrovascular disorders in SHRSP rats. It is an endogenous metabolite found in human milk. Studies have shown that a diet containing 1% Palmitoleic acid significantly improves the survival rate and reduces the incidence of stroke in SHRSP rats, even with excess NaCl intake.

  • Endogenous metabolite
  • Purity of 99.84%
  • Formula C16H30O2
  • CAS number 373-49-9
  • Appears as a colorless to light yellow liquid
  • Significant improvement in survival rate and reduced incidence of stroke in SHRSP rats

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.