Learn More
Selleck Chemical LLC Palmitoleic acid S3341-25mg
SDP
Catalog No. 503108573
Encompass_Preferred
Palmitoleic acid (POA Palmitoleate) stimulates the uptake of glucose in liver through activation of AMPK and FGF-21 dependent on PPAR
Catalog No. 50-310-8573 Supplier Selleck Chemical LLC Supplier No. S334125MG
Palmitoleic acid (POA Palmitoleate) stimulates the uptake of glucose in liver through activation of AMPK and FGF-21 dependent on PPAR