Learn More
TARGETMOL CHEMICALS INC Palmitoleic acid 500MG
SDP
Catalog No. 502878940
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Palmitoleic acid ((Z)-Hexadec-9-enoic acid) is found to be associated with isovaleric acidemia, which is an inborn error of metabolism. Purity 99.31%
Catalog No. 50-287-8940 Supplier TARGETMOL CHEMICALS INC Supplier No. T4872500MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Palmitoleic acid ((Z)-Hexadec-9-enoic acid) is found to be associated with isovaleric acidemia, which is an inborn error of metabolism. Purity 99.31%