Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitoleic acid, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018625 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8625 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8625 Supplier MilliporeSigma Supelco Supplier No. 761691G
Add to Cart
Add to Cart

Palmitoleic acid is a monounsaturated fatty acid found in macadamia oil and also in blood plasma. In the human body it is produced via endogenous fat synthesis.

TRUSTED_SUSTAINABILITY

Specifications

CAS 373-49-9
Density 0.895 g/mL (at 20°C)
Flash Point 62°C - Closed Cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)5 CH=CH(CH2)7 COOH
MDL Number MFCD00004437
Melting Point 0.5°C
Boiling Point 162°C (0.6 mmHg)
Synonym cis-9-Hexadecenoic acid
Type Analytical Standard
Quantity 1 g
Formula Weight 254.41
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.