Learn More
Cayman Chemical PalmItoleIc AcId-d13 25mg
SDP
Catalog No. 502941389
Encompass_Preferred
An internal standard for the quantification of palmitoleic acid by GC- or LC-MS
Catalog No. 50-294-1389 Supplier Cayman Chemical Supplier No. 2771425 MG
An internal standard for the quantification of palmitoleic acid by GC- or LC-MS