Learn More
Medchemexpress LLC Palmitoleic acid-d13 | 2692623-92-8 | 99.9% | 267.49 | 1 MG
SDP
Catalog No. 5000453708
Encompass_Preferred
Palmitoleic acid-d13 is a deuterium-labeled form of Palmitoleic acid. Palmitoleic acid, a fatty acid, has been shown to prevent death from cerebrovascular disorders in SHRSP rats.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for use as tracers in drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-53708 Supplier Medchemexpress LLC Supplier No. HYW011873S11MG
Palmitoleic acid-d13 is a deuterium-labeled form of Palmitoleic acid. Palmitoleic acid, a fatty acid, has been shown to prevent death from cerebrovascular disorders in SHRSP rats.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for use as tracers in drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.