Learn More
Cayman Chemical Palmitoleic Acid-d13, Purity: ≥99% deuterated forms (d1-d13), CAS: 2692623-92-8, 5 mg
SDP
Catalog No. 502937755
Encompass_Preferred
An internal standard for the quantification of palmitoleic acid by GC- or LC-MS
Catalog No. 50-293-7755 Supplier Cayman Chemical Supplier No. 277145 MG
An internal standard for the quantification of palmitoleic acid by GC- or LC-MS