Learn More
Cayman Chemical Palmitoleoyl 3carbacyclic Pho
SDP
Supplier:  Cayman Chemical 1001029810 MG
Encompass_Preferred
A cyclic LPA analog that contains the 16:1 fatty acid, palmitoleate, at the sn-1 position; at 25 μM inhibits the transcellular migration of MM1 cells across mesothelial cell monolayers without affecting proliferation; significantly inhibits autotaxin (IC50 = 620 nM)
Catalog No. 50-288-8493