Learn More
Medchemexpress LLC Palmitoleoyl chlorid 100mg | 40426-22-0 | 272.85 | C16H29ClO | 100 MG
SDP
Catalog No. 5000227286
Encompass_Preferred
Palmitoleoyl chloride is an acyl chloride derivative of palmitoleic acid used as a reactive intermediate in organic synthesis. Supplied as a liquid reagent for research applications, it is employed to introduce the palmitoleoyl acyl group in acylation and coupling reactions.
  • Reactive acyl chloride for acylation chemistry.
  • Useful intermediate for synthesis of lipid-derived and small-molecule compounds.
  • Available in lab-scale quantities, including 100 MG packaging.
  • Typical purity around 95% as supplied.
  • Requires low-temperature storage to preserve stability and shelf life.
Catalog No. 50-002-27286 Supplier Medchemexpress LLC Supplier No. HY116955100MG
Palmitoleoyl chloride is an acyl chloride derivative of palmitoleic acid used as a reactive intermediate in organic synthesis. Supplied as a liquid reagent for research applications, it is employed to introduce the palmitoleoyl acyl group in acylation and coupling reactions.
  • Reactive acyl chloride for acylation chemistry.
  • Useful intermediate for synthesis of lipid-derived and small-molecule compounds.
  • Available in lab-scale quantities, including 100 MG packaging.
  • Typical purity around 95% as supplied.
  • Requires low-temperature storage to preserve stability and shelf life.