Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitoleoyl chlorid 50mg | 40426-22-0 | 272.85 g/mol | C16H29ClO | 50 MG
SDP

Catalog No. 5000227190
Encompass_Preferred

Palmitoleoyl chloride is an acyl chloride derivative of palmitoleic acid used as a reactive intermediate in organic and medicinal chemistry. Supplied for research use, it is commonly used to introduce a palmitoleoyl (C16:1) acyl group into target molecules.

  • Purity 95.0%.
  • Chemical formula C16H29ClO; molecular weight 272.85 g/mol.
  • Appearance: light yellow to brown liquid.
  • Soluble in DMSO (≈50 mg/mL; may require ultrasonic).
  • Storage: recommended -20°C for long-term storage; additional guidance for solutions applies.
  • Intended for research use only.

Catalog No. 50-002-27190 Supplier Medchemexpress LLC Supplier No. HY11695550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitoleoyl chloride is an acyl chloride derivative of palmitoleic acid used as a reactive intermediate in organic and medicinal chemistry. Supplied for research use, it is commonly used to introduce a palmitoleoyl (C16:1) acyl group into target molecules.

  • Purity 95.0%.
  • Chemical formula C16H29ClO; molecular weight 272.85 g/mol.
  • Appearance: light yellow to brown liquid.
  • Soluble in DMSO (≈50 mg/mL; may require ultrasonic).
  • Storage: recommended -20°C for long-term storage; additional guidance for solutions applies.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.