Learn More
Medchemexpress LLC Palmitoleoyl-CoA | 18198-76-0 | 93.0% | 1003.93 | C37H64N7O17P3S | 5 MG
SDP
Catalog No. 5000222013
Encompass_Preferred
Palmitoleoyl-CoA is a fatty acyl-coenzyme A reagent used in biochemical and metabolic studies. It participates in mitochondrial β-oxidation, can induce mitochondrial membrane permeability transition, and modulates metabolism via allosteric control of AMPK β1 isoforms, making it useful for assays investigating lipid metabolism and mitochondrial function.
  • High purity (93.0%) suitable for biochemical assays.
  • Useful for studies of mitochondrial β-oxidation and membrane permeability.
  • Modulates AMPK β1 isoforms for metabolism-related experiments.
  • Supplied in a 5 mg pack for small-scale assays.
  • Molecular weight 1003.93 and formula C37H64N7O17P3S for reference.
Catalog No. 50-002-22013 Supplier Medchemexpress LLC Supplier No. HY1377825MG
Palmitoleoyl-CoA is a fatty acyl-coenzyme A reagent used in biochemical and metabolic studies. It participates in mitochondrial β-oxidation, can induce mitochondrial membrane permeability transition, and modulates metabolism via allosteric control of AMPK β1 isoforms, making it useful for assays investigating lipid metabolism and mitochondrial function.
  • High purity (93.0%) suitable for biochemical assays.
  • Useful for studies of mitochondrial β-oxidation and membrane permeability.
  • Modulates AMPK β1 isoforms for metabolism-related experiments.
  • Supplied in a 5 mg pack for small-scale assays.
  • Molecular weight 1003.93 and formula C37H64N7O17P3S for reference.