Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitoleoyl ethanolamide | 94421-67-7 | 99.8% | 10 MG
SDP

Catalog No. 5000340037
Encompass_Preferred

Palmitoleoyl ethanolamide (POEA) is an endogenous fatty amide that exhibits biological activity. It is provided as a solid, white to off-white in color, with a molecular weight of 297.48 and the chemical formula C18H35NO2.

  • Endogenous fatty amide
  • Solid, white to off-white appearance
  • Molecular weight: 297.48
  • Chemical formula: C18H35NO2
  • Purity: 99.77%
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-003-40037 Supplier Medchemexpress LLC Supplier No. HY12706110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitoleoyl ethanolamide (POEA) is an endogenous fatty amide that exhibits biological activity. It is provided as a solid, white to off-white in color, with a molecular weight of 297.48 and the chemical formula C18H35NO2.

  • Endogenous fatty amide
  • Solid, white to off-white appearance
  • Molecular weight: 297.48
  • Chemical formula: C18H35NO2
  • Purity: 99.77%
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.