Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitoleoyl ethanolamide | 94421-67-7 | MFCD30562006 | 99.8% | 297.48 g/mol | C18H35NO2 | 5 MG
SDP

Catalog No. 5000264460
Encompass_Preferred

Palmitoleoyl ethanolamide is an endogenous fatty amide supplied for research use, reported to act in lipid signaling and inflammatory pathways. It is provided as a white to off-white solid with high reported purity and includes solubility and storage guidance for both in vitro and in vivo applications.

  • Endogenous fatty amide with biological activity.
  • High purity (reported 99.77%).
  • White to off-white solid for laboratory use.
  • Soluble in DMSO; in vivo formulation protocols available.
  • Recommended storage: powder at -20°C or 4°C; solvent storage guidance provided.
  • Offered in small research pack sizes suitable for preclinical studies.

Catalog No. 50-002-64460 Supplier Medchemexpress LLC Supplier No. HY1270615MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitoleoyl ethanolamide is an endogenous fatty amide supplied for research use, reported to act in lipid signaling and inflammatory pathways. It is provided as a white to off-white solid with high reported purity and includes solubility and storage guidance for both in vitro and in vivo applications.

  • Endogenous fatty amide with biological activity.
  • High purity (reported 99.77%).
  • White to off-white solid for laboratory use.
  • Soluble in DMSO; in vivo formulation protocols available.
  • Recommended storage: powder at -20°C or 4°C; solvent storage guidance provided.
  • Offered in small research pack sizes suitable for preclinical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.