Learn More
Cayman Chemical Palmitoyl 3carbacyclic Phosph
SDP
Supplier:  Cayman Chemical 100102931 MG
Encompass_Preferred
Contains the 16:0 fatty acid, palmitate, at the sn-1 position; at 25 μM inhibits the transcellular migration of MM1 cells across mesothelial cell monolayers without affecting proliferation; at 0.1-25 μM significantly inhibits autotaxin
Catalog No. 50-289-1275