Learn More
Apexbio Technology LLC PALMITOYL-CARNITINE-D3-HCL-1MG
SDP
Catalog No. 5000570729
Encompass_Preferred
Palmitoyl carnitine-d3 HCl (CAS No 1334532-26-1) is a deuterated derivative of palmitoyl carnitine often used in biomedical research focusing on lipid metabolism and cellular energy processes This compound is involved in the transport of long-chain fatty acids into the mitochondria thereby influencing beta-oxidation pathways Its activity is typically observed in the nanomolar to low micromolar range making it suitable for precise biochemical assays Palmitoyl carnitine-d3 HCl is frequently utilized in various in vitro models to investigate metabolic functions as well as in animal studies for elucidating mitochondrial dynamics and energy regulation It also finds applications in drug screening processes to assess effects on lipid metabolism pathways Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70729 Supplier Apexbio Technology LLC Supplier No. SL10101
Palmitoyl carnitine-d3 HCl (CAS No 1334532-26-1) is a deuterated derivative of palmitoyl carnitine often used in biomedical research focusing on lipid metabolism and cellular energy processes This compound is involved in the transport of long-chain fatty acids into the mitochondria thereby influencing beta-oxidation pathways Its activity is typically observed in the nanomolar to low micromolar range making it suitable for precise biochemical assays Palmitoyl carnitine-d3 HCl is frequently utilized in various in vitro models to investigate metabolic functions as well as in animal studies for elucidating mitochondrial dynamics and energy regulation It also finds applications in drug screening processes to assess effects on lipid metabolism pathways Experimental concentrations typically depend on the specific research design