Learn More
Cayman Chemical Palmitoyl Ethanolamided4
SDP
Catalog No. 502889279
Encompass_Preferred
An internal standard for the quantification of palmitoyl ethanolamide by GC- or LC-MS
Catalog No. 50-288-9279 Supplier Cayman Chemical Supplier No. 10007824100 UG
An internal standard for the quantification of palmitoyl ethanolamide by GC- or LC-MS