Learn More
Cayman Chemical Palmitoyl Ethanolamided4
SDP
Supplier:  Cayman Chemical 100078241 MG
Encompass_Preferred
An internal standard for the quantification of palmitoyl ethanolamide by GC- or LC-MS
Catalog No. 50-288-8174