Learn More
Cayman Chemical PalmItoyl EthnolamIde-d4 100UG
SDP
Catalog No. 502892895
Encompass_Preferred
An internal standard for the quantification of palmitoyl ethanolamide by GC- or LC-MS
Catalog No. 50-289-2895 Supplier Cayman Chemical Supplier No. 9000551100 UG
An internal standard for the quantification of palmitoyl ethanolamide by GC- or LC-MS