Learn More
Cayman Chemical Palmitoyl Ethanolamide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 946524-34-1, 1 mg
SDP
Catalog No. 502894528
Encompass_Preferred
An internal standard for the quantification of palmitoyl ethanolamide by GC- or LC-MS
Catalog No. 50-289-4528 Supplier Cayman Chemical Supplier No. 90005511 MG
An internal standard for the quantification of palmitoyl ethanolamide by GC- or LC-MS