Learn More
Cayman Chemical PalmItoyl EthnolamIde-d4 5mg
SDP
Catalog No. 502893675
Encompass_Preferred
An internal standard for the quantification of palmitoyl ethanolamide by GC- or LC-MS
Catalog No. 50-289-3675 Supplier Cayman Chemical Supplier No. 90005515 MG
An internal standard for the quantification of palmitoyl ethanolamide by GC- or LC-MS