Learn More
Medchemexpress LLC Palmitoyl hexapeptide-12 | 171263-26-6 | 97.4% | C38H68N6O8 | 100 MG
SDP
Catalog No. 5000434248
Encompass_Preferred
Palmitoyl hexapeptide-12 is a bioactive peptide that reduces IL-6 and exhibits anti-aging effects. It can be used in research related to photoaging and age-related skin problems.
  • Reduces IL-6.
  • Has anti-aging effects.
  • Useful in research for photoaging and related aging skin problems.
Catalog No. 50-004-34248 Supplier Medchemexpress LLC Supplier No. HY66015100MG
Palmitoyl hexapeptide-12 is a bioactive peptide that reduces IL-6 and exhibits anti-aging effects. It can be used in research related to photoaging and age-related skin problems.
  • Reduces IL-6.
  • Has anti-aging effects.
  • Useful in research for photoaging and related aging skin problems.