Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitoyl-L-carnitine, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013637 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3637 10 mg
11-101-3638 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3637 Supplier MilliporeSigma Supelco Supplier No. 9150310MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 2364-67-2
Linear Formula C23H45 NO4
Synonym (2 R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexadecyl)oxy]-1-propanaminium inner salt; L-Carnitine hexadecanoyl ester; C16-Carnitine; Hexadecanoyl-L-carnitine
Type Analytical Standard
Quantity 10 mg
Formula Weight 399.61
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.