Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitoyl-L-carnitine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. P254825MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P254825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P254825MG Supplier TCI America Supplier No. P254825MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18877-64-0
Molecular Formula C23H46ClNO4
Molecular Weight (g/mol) 436.074
MDL Number MFCD00070346
InChI Key GAMKNLFIHBMGQT-ZMBIFBSDSA-N
Synonym (R)-3-(Hexadecanoyloxy)-4-(trimethylammonio)butyrate Hydrochloride, (R)-3-(Palmitoyloxy)-4-(trimethylammonio)butanoate Hydrochloride
PubChem CID 167759
IUPAC Name [(2R)-3-carboxy-2-hexadecanoyloxypropyl]-trimethylazanium;chloride
SMILES CCCCCCCCCCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]

Specifications

Melting Point 178°C
Color White
Quantity 25 mg
Formula Weight 436.07
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Palmitoyl-L-carnitine Hydrochloride
EINECSNumber (2)-2756
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.