Learn More
Medchemexpress LLC Palmitoyl pentapeptide 4 | 214047-00-4 | 99.7% | 802.05 | 100 MG
SDP
Catalog No. 5000425050
Encompass_Preferred
Palmitoyl pentapeptide 4 is a bioactive peptide with anti-ageing effects, commonly used as a cosmetic ingredient. It acts as a signal peptide that promotes collagen synthesis by stimulating fibroblasts to produce collagen (types I, III, and IV), fibronectin, and glycosaminoglycans. This makes it a safe and effective anti-aging ingredient for skin improvement.
  • Promotes collagen synthesis
  • Improves skin elasticity
  • Reduces wrinkles
  • Enhances skin barrier function
Catalog No. 50-004-25050 Supplier Medchemexpress LLC Supplier No. HYP0045100MG
Palmitoyl pentapeptide 4 is a bioactive peptide with anti-ageing effects, commonly used as a cosmetic ingredient. It acts as a signal peptide that promotes collagen synthesis by stimulating fibroblasts to produce collagen (types I, III, and IV), fibronectin, and glycosaminoglycans. This makes it a safe and effective anti-aging ingredient for skin improvement.
  • Promotes collagen synthesis
  • Improves skin elasticity
  • Reduces wrinkles
  • Enhances skin barrier function