Learn More
Medchemexpress LLC Palmitoyl pentapeptide 4 | 214047-00-4 | 99.7% | 802.05 | 250 MG
SDP
Catalog No. 5000426948
Encompass_Preferred
Palmitoyl pentapeptide 4 is a bioactive peptide with anti-ageing effects, commonly used as a cosmetic ingredient. It acts as a signal peptide, promoting collagen synthesis within the skin.
  • Stimulates fibroblasts to produce collagen (types I, III, and IV), fibronectin, and glycosaminoglycans.
  • Safe and effective anti-aging ingredient.
  • Improves skin elasticity.
  • Reduces wrinkles.
  • Improves skin barrier function.
Catalog No. 50-004-26948 Supplier Medchemexpress LLC Supplier No. 250MGHYP0045
Palmitoyl pentapeptide 4 is a bioactive peptide with anti-ageing effects, commonly used as a cosmetic ingredient. It acts as a signal peptide, promoting collagen synthesis within the skin.
  • Stimulates fibroblasts to produce collagen (types I, III, and IV), fibronectin, and glycosaminoglycans.
  • Safe and effective anti-aging ingredient.
  • Improves skin elasticity.
  • Reduces wrinkles.
  • Improves skin barrier function.