Learn More
Cayman Chemical Palmitoyl Serotonin, Purity: ≥98%, CAS: 212707-51-2, 10 mg
SDP
Catalog No. 502894923
Encompass_Preferred
A hybrid molecule patterned after N-arachidonoyl serotonin; while saturated 11- or 12-carbon fatty acids linked to serotonin potently inhibit capsaicin-induced TRPV1 channel activation (IC50 = 0.76 μM) without blocking FAAH-mediated hydrolysis of arachidonoyl ethanolamine (IC50 > 50 μM), the effects of palmitoyl serotonin are unknown
Catalog No. 50-289-4923 Supplier Cayman Chemical Supplier No. 900063010 MG
A hybrid molecule patterned after N-arachidonoyl serotonin; while saturated 11- or 12-carbon fatty acids linked to serotonin potently inhibit capsaicin-induced TRPV1 channel activation (IC50 = 0.76 μM) without blocking FAAH-mediated hydrolysis of arachidonoyl ethanolamine (IC50 > 50 μM), the effects of palmitoyl serotonin are unknown