Learn More
Medchemexpress LLC Palmitoyl serotonin | 212707-51-2 | 99.0% | C26H42N2O2 | 10MG
SDP
Catalog No. 5000261143
Encompass_Preferred
Palmitoyl serotonin is a research-grade small molecule used as a biochemical probe of fatty acid amide hydrolase (FAAH)-related pathways. Supplied as a purified solid (99.0% purity), it is employed in preclinical studies investigating motor effects and Parkinson's disease models. Not for human consumption.
  • High purity suitable for biochemical assays and preclinical studies.
  • Confirmed molecular formula C26H42N2O2 and molecular weight 414.62.
  • Available in multiple small-scale quantities for research workflows.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet.
  • Useful as a probe in FAAH and motor dysfunction research models.
Catalog No. 50-002-61143 Supplier Medchemexpress LLC Supplier No. HY13167810MG
Palmitoyl serotonin is a research-grade small molecule used as a biochemical probe of fatty acid amide hydrolase (FAAH)-related pathways. Supplied as a purified solid (99.0% purity), it is employed in preclinical studies investigating motor effects and Parkinson's disease models. Not for human consumption.
  • High purity suitable for biochemical assays and preclinical studies.
  • Confirmed molecular formula C26H42N2O2 and molecular weight 414.62.
  • Available in multiple small-scale quantities for research workflows.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet.
  • Useful as a probe in FAAH and motor dysfunction research models.