Learn More
Cayman Chemical PalmItoyl SerotonIn 5mg
SDP
Catalog No. 502893512
Encompass_Preferred
A hybrid molecule patterned after N-arachidonoyl serotonin; while saturated 11- or 12-carbon fatty acids linked to serotonin potently inhibit capsaicin-induced TRPV1 channel activation (IC50 = 0.76 μM) without blocking FAAH-mediated hydrolysis of arachidonoyl ethanolamine (IC50 > 50 μM), the effects of palmitoyl serotonin are unknown
Catalog No. 50-289-3512 Supplier Cayman Chemical Supplier No. 90006305 MG
Add to Cart
Add to Cart
A hybrid molecule patterned after N-arachidonoyl serotonin; while saturated 11- or 12-carbon fatty acids linked to serotonin potently inhibit capsaicin-induced TRPV1 channel activation (IC50 = 0.76 μM) without blocking FAAH-mediated hydrolysis of arachidonoyl ethanolamine (IC50 > 50 μM), the effects of palmitoyl serotonin are unknown