Learn More
Medchemexpress LLC Palmitoyl tetrapeptide-10 | 887140-79-6 | 99.5% | 761.05 | 10 MG
SDP
Catalog No. 5000396366
Encompass_Preferred
Palmitoyl tetrapeptide-10 is a bioactive peptide that works synergistically with other active ingredients to increase the expression of corneodesmosin (a marker of keratinocyte adhesion) and filaggrin (a marker of keratinocyte terminal differentiation). This action helps in reducing skin permeability and improving skin barrier function. It can be used in the research of sensitive skin syndrome and skin pigmentation.
  • Increases expression of corneodesmosin
  • Increases expression of filaggrin
  • Reduces skin permeability
  • Improves skin barrier function
  • Suitable for research on sensitive skin syndrome
  • Suitable for research on skin pigmentation
Catalog No. 50-003-96366 Supplier Medchemexpress LLC Supplier No. HYP526910MG
Palmitoyl tetrapeptide-10 is a bioactive peptide that works synergistically with other active ingredients to increase the expression of corneodesmosin (a marker of keratinocyte adhesion) and filaggrin (a marker of keratinocyte terminal differentiation). This action helps in reducing skin permeability and improving skin barrier function. It can be used in the research of sensitive skin syndrome and skin pigmentation.
  • Increases expression of corneodesmosin
  • Increases expression of filaggrin
  • Reduces skin permeability
  • Improves skin barrier function
  • Suitable for research on sensitive skin syndrome
  • Suitable for research on skin pigmentation