Learn More
Medchemexpress LLC Palmitoyl tetrapeptide-20 | 99.21% | 739.00 | 10 MG
SDP
Catalog No. 5000396367
Encompass_Preferred
Palmitoyl tetrapeptide-20 (PTP20) is a biomimetic peptide agonist of α-MSH. It promotes hair pigmentation and delays hair graying by activating the MC1-R pathway, enhancing catalase activity to reduce H2O2 accumulation, and upregulating SIRT1 activity. This product can be used in research on preventing hair loss and improving hair graying.
  • Promotes hair pigmentation
  • Delays hair graying
  • Activates MC1-R pathway
  • Enhances catalase activity
  • Reduces H2O2 accumulation
  • Upregulates SIRT1 activity
  • Suitable for research on preventing hair loss
  • Useful for improving hair graying
Catalog No. 50-003-96367 Supplier Medchemexpress LLC Supplier No. HYP524710MG
Palmitoyl tetrapeptide-20 (PTP20) is a biomimetic peptide agonist of α-MSH. It promotes hair pigmentation and delays hair graying by activating the MC1-R pathway, enhancing catalase activity to reduce H2O2 accumulation, and upregulating SIRT1 activity. This product can be used in research on preventing hair loss and improving hair graying.
  • Promotes hair pigmentation
  • Delays hair graying
  • Activates MC1-R pathway
  • Enhances catalase activity
  • Reduces H2O2 accumulation
  • Upregulates SIRT1 activity
  • Suitable for research on preventing hair loss
  • Useful for improving hair graying