Learn More
Medchemexpress LLC Palmitoyl Tetrapeptide-3 | 221227-05-0 | 99.2% | 694.91 | 100 MG
SDP
Catalog No. 5000426619
Encompass_Preferred
Palmitoyl Tetrapeptide-3 (Palmitoyl Tetrapeptide-7) is a synthetic peptide corresponding to amino acids 341-344 of the human immunoglobulin heavy chain, capable of stimulating phagocytosis. It offers several benefits:
  • Reduces interleukin-6 (IL-6) secretion in keratinocytes.
  • Inhibits the UVB radiation-exposure inflammatory response of the skin.
  • Possesses anti-inflammatory and anti-aging effects.
  • Contributes to the reduction of skin wrinkles by promoting the production of elastic fibers in the papillary dermis.
Catalog No. 50-004-26619 Supplier Medchemexpress LLC Supplier No. HYP0064100MG
Palmitoyl Tetrapeptide-3 (Palmitoyl Tetrapeptide-7) is a synthetic peptide corresponding to amino acids 341-344 of the human immunoglobulin heavy chain, capable of stimulating phagocytosis. It offers several benefits:
  • Reduces interleukin-6 (IL-6) secretion in keratinocytes.
  • Inhibits the UVB radiation-exposure inflammatory response of the skin.
  • Possesses anti-inflammatory and anti-aging effects.
  • Contributes to the reduction of skin wrinkles by promoting the production of elastic fibers in the papillary dermis.