Learn More
Cayman Chemical Palmitoyl thioPC
SDP
Supplier:  Cayman Chemical 100105211 MG
Encompass_Preferred
A chromogenic substrate for PLA2; hydrolysis of the thioester by PLA2 yields a free thiol that reacts readily with DTNB with an absorbance maximum at 412 nm; has been used to measure bee venom sPLA2 activity in a phospholipid:Triton X-100 mixed micelle system
Catalog No. 50-288-9696