Learn More
Apexbio Technology LLC PalMitoyl Tripeptide-1 147732-56-7 1g
SDP
Catalog No. 5000090173
Encompass_Preferred
Palmitoyl Tripeptide-1 is a synthetic peptide derivative designed to modulate extracellular matrix (ECM) remodeling by interacting with dermal fibroblast cells It facilitates its biological effects by binding cellular receptors and stimulating intracellular signaling pathways involved in collagen synthesis and ECM reorganization Palmitoyl Tripeptide-1 exerts its biological activity primarily through the promotion of collagen synthesis and modulation of fibroblast biochemical activity Based on these pharmacological properties Palmitoyl Tripeptide-1 holds research potential in studies of skin aging dermal regeneration ECM homeostasis and cellular response dynamics mediated by bioactive peptides
Catalog No. 50-000-90173 Supplier Apexbio Technology LLC Supplier No. B60531000
Palmitoyl Tripeptide-1 is a synthetic peptide derivative designed to modulate extracellular matrix (ECM) remodeling by interacting with dermal fibroblast cells It facilitates its biological effects by binding cellular receptors and stimulating intracellular signaling pathways involved in collagen synthesis and ECM reorganization Palmitoyl Tripeptide-1 exerts its biological activity primarily through the promotion of collagen synthesis and modulation of fibroblast biochemical activity Based on these pharmacological properties Palmitoyl Tripeptide-1 holds research potential in studies of skin aging dermal regeneration ECM homeostasis and cellular response dynamics mediated by bioactive peptides