Learn More
Medchemexpress LLC Palmitoyl Tripeptide-1 | 147732-56-7 | 99.2% | 578.79 | 250 MG
SDP
Catalog No. 5000426236
Encompass_Preferred
Palmitoyl tripeptide-1 is a fatty acid modified polypeptide known for its wrinkle-removing effects. It is involved in the feedback regulation and control of connective tissue reconstruction and cell proliferation, and can promote the generation of skin repair protein.
  • Performs feedback regulation and control on connective tissue reconstruction and cell proliferation.
  • Promotes the generation of skin repair protein.
Catalog No. 50-004-26236 Supplier Medchemexpress LLC Supplier No. 250MGHYP0047A
Add to Cart
Add to Cart
Palmitoyl tripeptide-1 is a fatty acid modified polypeptide known for its wrinkle-removing effects. It is involved in the feedback regulation and control of connective tissue reconstruction and cell proliferation, and can promote the generation of skin repair protein.
  • Performs feedback regulation and control on connective tissue reconstruction and cell proliferation.
  • Promotes the generation of skin repair protein.