Learn More
Medchemexpress LLC Palmitoyl Tripeptide-1 | 147732-56-7 | 99.23% | 578.79 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP0047A100MG
Encompass_Preferred
Palmitoyl Tripeptide-1 is a fatty acid modified polypeptide known for its significant wrinkle-removing effects. This bioactive peptide actively regulates and controls the processes involved in connective tissue reconstruction and cell proliferation, essential for skin repair.
  • Promotes the generation of skin repair protein.
  • Performs feedback regulation on connective tissue reconstruction.
  • Controls cell proliferation for skin health.
  • Suitable for research applications.
Catalog No. 50-004-26519