Learn More
Medchemexpress LLC Palmitoylcarnitine | 1935-18-8 | MFCD00152152 | 100.0% | 399.61 g/mol | C23H45NO4 | 5MG
SDP
Catalog No. 5000235410
Encompass_Preferred
Palmitoylcarnitine is an endogenous O-acylcarnitine metabolite used as a research reagent and reference standard in studies of fatty acid metabolism and mitochondrial function. It is supplied as a solid powder with high stated purity and recommended low-temperature storage for long-term stability.
  • High purity suitable for analytical applications.
  • Solid powder form for easy handling and weighing.
  • Available in small research sizes, including 5 mg.
  • Stable when stored at low temperatures for extended periods.
  • Provides a standard for metabolic and biochemical assays.
Catalog No. 50-002-35410 Supplier Medchemexpress LLC Supplier No. HY1263575MG
Palmitoylcarnitine is an endogenous O-acylcarnitine metabolite used as a research reagent and reference standard in studies of fatty acid metabolism and mitochondrial function. It is supplied as a solid powder with high stated purity and recommended low-temperature storage for long-term stability.
  • High purity suitable for analytical applications.
  • Solid powder form for easy handling and weighing.
  • Available in small research sizes, including 5 mg.
  • Stable when stored at low temperatures for extended periods.
  • Provides a standard for metabolic and biochemical assays.