Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitoylcarnitine chloride | 6865-14-1 | MFCD00063481 | 98.0% | 436.07 | C23H46ClNO4 | 25 MG
SDP

Catalog No. 5000212845
Encompass_Preferred

Palmitoylcarnitine chloride is a fatty-acid derived mitochondrial substrate used in biochemical and cell biology research. It modulates Ca2+ influx and pro-inflammatory signaling and has been reported to reduce cell viability in certain colorectal and prostate cancer models. Supplied as a solid reagent with recommended sealed storage at -20°C.

  • Fatty-acid derived mitochondrial substrate for metabolic studies.
  • Modulates Ca2+ influx and pro-inflammatory signaling pathways.
  • Reported to decrease cell survival in specific cancer cell models.
  • High reported purity: 98.0% (NMR).
  • Molecular formula C23H46ClNO4; molecular weight 436.07.
  • Recommended storage: -20°C, sealed, away from moisture.

Catalog No. 50-002-12845 Supplier Medchemexpress LLC Supplier No. HY10101725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitoylcarnitine chloride is a fatty-acid derived mitochondrial substrate used in biochemical and cell biology research. It modulates Ca2+ influx and pro-inflammatory signaling and has been reported to reduce cell viability in certain colorectal and prostate cancer models. Supplied as a solid reagent with recommended sealed storage at -20°C.

  • Fatty-acid derived mitochondrial substrate for metabolic studies.
  • Modulates Ca2+ influx and pro-inflammatory signaling pathways.
  • Reported to decrease cell survival in specific cancer cell models.
  • High reported purity: 98.0% (NMR).
  • Molecular formula C23H46ClNO4; molecular weight 436.07.
  • Recommended storage: -20°C, sealed, away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.