Learn More
Cayman Chemical Palmitoyl-DL-carnitine (chloride), Purity: ≥95%, CAS: 6865-14-1, 100 mg
SDP
Catalog No. 502951351
Encompass_Preferred
A long-chain acylcarnitine with both intracellular and extracellular roles; transported into mitochondria to deliver palmitate for fatty acid oxidation; alters calcium mobilization in many cell types; also affects endothelial and epithelial tight junctions, reducing resistance and increasing permeability
Catalog No. 50-295-1351 Supplier Cayman Chemical Supplier No. 11095100 MG
A long-chain acylcarnitine with both intracellular and extracellular roles; transported into mitochondria to deliver palmitate for fatty acid oxidation; alters calcium mobilization in many cell types; also affects endothelial and epithelial tight junctions, reducing resistance and increasing permeability