Learn More
Medchemexpress LLC Palmitoylethanolamide | 544-31-0 | MFCD00020562 | 99.86% | 299.49 | 200 MG
SDP
Catalog No. 5000401005
Encompass_Preferred
Palmitoylethanolamide (Standard) is the analytical standard for Palmitoylethanolamide, an active endogenous compound. It is intended for research and analytical applications, and can be used for preventing virus infection of the respiratory tract.
  • Analytical standard for Palmitoylethanolamide.
  • Intended for research and analytical applications.
  • Can be used for preventing virus infection of the respiratory tract.
  • High purity of 99.86%.
  • Available in multiple quantities for various research needs.
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-01005 Supplier Medchemexpress LLC Supplier No. HY20685200MG
Palmitoylethanolamide (Standard) is the analytical standard for Palmitoylethanolamide, an active endogenous compound. It is intended for research and analytical applications, and can be used for preventing virus infection of the respiratory tract.
  • Analytical standard for Palmitoylethanolamide.
  • Intended for research and analytical applications.
  • Can be used for preventing virus infection of the respiratory tract.
  • High purity of 99.86%.
  • Available in multiple quantities for various research needs.
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.