Learn More
Cayman Chemical Palmitoyl-L-carnitine (chloride), Purity: ≥95%, CAS: 18877-64-0, 50 mg
SDP
Catalog No. 502935464
Encompass_Preferred
A long-chain acylcarnitine; delivers palmitate into the mitochondria for fatty acid oxidation and energy production; inhibits lecithin:cholesterol acyltransferase activity in isolated rat plasma at 500 µM; serum and hepatic levels are increased in mice during cold exposure; protects against age-induced cold sensitivity in mice
Catalog No. 50-293-5464 Supplier Cayman Chemical Supplier No. 2655350 MG
A long-chain acylcarnitine; delivers palmitate into the mitochondria for fatty acid oxidation and energy production; inhibits lecithin:cholesterol acyltransferase activity in isolated rat plasma at 500 µM; serum and hepatic levels are increased in mice during cold exposure; protects against age-induced cold sensitivity in mice