Learn More
Enzo Life Sciences Palmitylethanolamide (50mg). CAS: 544-31-0
SDP
Supplier:  Enzo Life Sciences BMLFA0180050
Encompass_Preferred
Endogenous cannabinoid. Weak ligand of CB1 (Ki=23.8µM) and CB2 (Ki=13.9µM) receptor. Inhibits fatty acid amide hydrolase (FAAH) (IC50=5.1µM). Immunosuppressant, anti-inflammatory, anti-nociceptive and anti-convulsant in vivo. The exact mode of action has not yet been revealed. It has been suggested that PEA: i) binds to a yet to be discovered cannabinoid receptor similar to CB2; ii) administered in vivo elicits the synthesis of endogenous agonists of CB2; iii) acts as an "entourage" compound by enhancing the activity and/or by influencing the turnover of endogenous agonists of CB2, possibly but not uniquely, by inhibiting their degradation. Alternative name: PEA, Palmidrol. Purity: ≥98% (TLC). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: +4°C.
Catalog No. 50-201-0805