Learn More
MSC Osborn Scouring Brush, Palmetto Fiber Bristles, 5 in Brush Length, Wood Handle
Catalog No. 502837389
Encompass
This scouring brush features durable palmetto fiber bristles and a sturdy wood handle, designed for effective utility work. It has a 5 inch brush length and an overall length of 21-3/4 inches, making it suitable for a variety of cleaning tasks.
  • Durable palmetto fiber bristles
  • 5 inch brush length
  • 21-3/4 inch overall length
  • Wood handle and block material
  • Designed for utility work applications
  • Oval brush head shape
  • Long handle style for extended reach
  • Brown bristle color
Catalog No. 50-283-7389 Supplier MSC Supplier No. 63439327
This scouring brush features durable palmetto fiber bristles and a sturdy wood handle, designed for effective utility work. It has a 5 inch brush length and an overall length of 21-3/4 inches, making it suitable for a variety of cleaning tasks.
  • Durable palmetto fiber bristles
  • 5 inch brush length
  • 21-3/4 inch overall length
  • Wood handle and block material
  • Designed for utility work applications
  • Oval brush head shape
  • Long handle style for extended reach
  • Brown bristle color